Learn More
Praseodymium(III, IV) oxide, 99.9% (REO)
Description
Praseodymium(III, IV) oxide is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2500°C |
| Physical Form | Sintered Lump |
| Quantity | 50 g |
| Solubility Information | Insoluble in water. Moderately soluble in strong mineral acids. |
| Formula Weight | 1021.43 |
| Percent Purity | 99.9% |
| Assay | (REO) |
| Chemical Name or Material | Praseodymium(III, IV) oxide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.