Learn More
Invitrogen™ Prasinezumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558431
 View more versions of this product
Product Code. 30283374
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Prasinezumab (PRX002/RG7935) is an investigational monoclonal antibody that targets alpha-synuclein, a protein that is believed to misfold and aggregate to form the protein structures that are highly implicated in Parkinson's disease pathology. Prasinezumab targets alpha-synuclein and is being developed through a worldwide collaboration with Roche as a potentially disease modifying therapy that is designed to slow or reduce the neurodegeneration associated with alpha-synuclein misfolding and/or its transmission.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Prasinezumab Humanized
Recombinant Monoclonal
Unconjugated
PRX002; RG-7935
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized