missing translation for 'onlineSavingsMsg'
Learn More

Prasugrel hydrochloride, MedChemExpress

Product Code. p-200097884
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451766 500 mg 500mg
16406716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451766 Supplier Medchem Express Supplier No. HY15284A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Prasugrel hydrochloride (PCR 4099 hydrochloride), a thienopyridine and prodrug, inhibits platelet function. Prasugrel hydrochloride is an orally active and potent P2Y12 receptor antagonist, and inhibits ADP-induced platelet aggregation.

Prasugrel hydrochloride (PCR 4099 hydrochloride), a thienopyridine and prodrug, inhibits platelet function. Prasugrel hydrochloride is an orally active and potent P2Y12 receptor antagonist, and inhibits ADP-induced platelet aggregation.

Specifications

Chemical Name or Material Prasugrel hydrochloride
CAS 389574-19-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H21ClFNO3S
Quantity 500 mg
Synonym PCR 4099 hydrochloride
Solubility Information DMSO : 41.67 mg/mL (101.66 mM; Need ultrasonic)
SMILES FC1=CC=CC=C1C(C(C2CC2)=O)N3CCC4=C(C3)C=C(S4)OC(C)=O.Cl
Molecular Weight (g/mol) 409.9
Formula Weight 409.9
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.