missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM4 Antibody (RAB-C367) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-20138-0.1mg
This item is not returnable.
View return policy
Description
PRDM4 Monoclonal antibody specifically detects PRDM4 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
Specifications
| PRDM4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Human | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay | |
| RAB-C367 | |
| Unconjugated | |
| MGC45046, PFM1PR domain-containing protein 4, PR domain containing 4, PR domain zinc finger protein 4, PR-domain zinc-finger protein PFM1 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM4 protein under non-denaturing conditions. | |
| 0.1 mg | |
| Chromatin Research | |
| 11108 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction