missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM5 Antibody (7D4C12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
203.00 € - 552.00 €
Specifications
| Antigen | PRDM5 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18157909
|
Novus Biologicals
NBP2-52540 |
0.1 mg |
552.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648217
|
Novus Biologicals
NBP2-52540-0.025mg |
0.025 mg |
203.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRDM5 Monoclonal specifically detects PRDM5 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| PRDM5 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| PFM2PR domain-containing protein 5, PR domain containing 5, PR domain zinc finger protein 5 | |
| PRDM5 | |
| IgG1 | |
| Protein A or G purified |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Chromatin Research, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 11107 | |
| Purified recombinant fragment of human PRDM5 (AA: 1-100) expressed in E. Coli. | |
| Primary | |
| 73 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title