Learn More
PRDM9 Antibody (RAB-C370) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20140-0.1mg
 View more versions of this product
Product Code. 18674354
  • 849.00 € / 0.10mg
Description
PRDM9 Monoclonal antibody specifically detects PRDM9 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRDM9
Monoclonal
1 mg/mL
PBS
Human
Affinity purified
RUO
56979
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay
RAB-C370
Unconjugated
EC 2.1.1.43, histone-lysine N-methyltransferase PRDM9, minisatellite binding protein 3 (115kD), minisatellite binding protein 3, 115kDa, PFM6MEISETZ, PR domain containing 9, PR domain zinc finger protein 9, PR domain-containing protein 9, PRMD9, ZNF899
This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM9 protein under non-denaturing conditions.
0.1 mg
Primary
Human
Purified