missing translation for 'onlineSavingsMsg'
Learn More

Pre-Mixed GFAA Matrix Modifier 1: 750 μg/mL Pd & 500 μg/mL Mg(NO3)2 in 2% HNO3, VHG™

Product Code. 30003163
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003163 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003163 Supplier VHG Labs Supplier No. VHGMPM1250
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Pre-Mixed GFAA Matrix Modifier 1: 750 μg/mL Pd & 500 μg/mL Mg(NO3)2 in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium Nitrate/Palladium
CAS 10377-60-3, 5-3-7440
Methods GFAA/AA
Analyte or Component Names Magnesium Nitrate/Palladium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 250 mL
Concentration or Composition (by Analyte or Components) Magnesium Nitrate 500 μg/mL/Palladium 750 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.