missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products Pre-Sterilized Whirl-Pak Bags
GREENER_CHOICE

Product Code. 30503529 Shop All Keystone Cleanroom Products Products
Click to view available options
No. per Pack:
20 Pk.
Unit Size:
Pack of 250
This item is not returnable. View return policy

Product Code. 30503529

Brand: Keystone Cleanroom Products S20319DPST10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-sterilized bags designed to reduce the risks of false positives of media plates

  • Designed to reduce the risks of false positives of media plates
  • This system provides complete protection from the time the plates are removed from the isolator all the way through incubation
  • The clear film material offers excellent clarity combined with strength and durability.
  • These bags feature the maximum protection against leaks with the continuous perimeter seal
  • Ready to use, double packed in easy to open Chevron-Peel DuPont™ Tyvek™ pouches, with an extra, sealed poly bag serving as a third sterile packaging layer
  • The inner and outer Tyvek™ iron Peel pouches feature hang holes that can be used to suspend the package inside the isolator for VHP decontamination
  • Validated sterile to a 10-6 SAL
  • Each Tyvek peel pouch package is labeled with an expiration date and a Lot number for complete traceability

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Certificate of Sterility
Color Clear
Disposable Single-use
For Use With (Application) Transport EM Media Plates
Material LDPE
Sterility Sterile
Latex Free Yes
No. per Pack 20 Pk.
Sterilization Method Gamma
Thickness (English) 3 mil
Length (Metric) 38.1 cm
Width (English) 7.5 in.
Width (Metric) 19.1 cm
Product Type Reclosable Bag
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.