Learn More
Uvex Preformed Mask silv-Air e 7312 FFP3
Product Code. 17532955 Shop All Uvex Products
Change view
Click to view available options
:
Filter Type: FFP3
Product Code.
17532955 Filter Type: FFP3
1 options
Product Code. 17532955
Brand: Uvex 8707312
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All masks of the silv-Air e family combine the unique technologies of optimum wearer comfort with extremely low breathing resistance. The products are therefore suitable for heavy work and for being worn for long periods of time.
General Features
  • Particle-filtering preformed mask with exhalation valve
  • Color: white
Protection Features
  • Certified FFP3 face mask to EN 149
  • D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • R label (reusable): tested for re-usability and performance, including in the second shift
Comfort Features
  • Soft material edges for increased wearer comfort
  • Very soft, all-around sealing lip for optimum wearer comfort and secure fit
  • Flexible, adjustable integrated nose clip
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (R marked, D marked)
Product Type Preformed Mask
With Exhalation Valve Yes
Filter Efficiency FFP3
Includes 360° exhalation valve, four-point head-securing strap, comfort nose clip, circumferential comfort sealing lip