Learn More
uvex Preformed Mask silv-Air p 7100 FFP1
Product Code. 17562945 Shop All Uvex Products
Change view
Click to view available options
Filter Type: FFP1
Product Code.
17562945 Filter Type: FFP1
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 17562945 Supplier uvex Supplier No. 8707100
  • ONLINE EXCLUSIVE PRICE
    38.95 €
    35.73 € / Pack of 20
    Save 3.22 €
    8% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
The gray coloring on the outside of the silv-Air p masks prevents them from looking dirty quickly and having to be disposed of prematurely by the user. By contrast, the inside is of course pure white. In addition and with all products in this family, the 3D nose geometry ensures an optimum fit, even with different face shapes.
General Features
  • Particle-filtering preformed mask
  • Metal-free processing
  • Soft material edges for increased wearer comfort
  • Seamless headband for a comfortable, secure fit
  • Color: silver-gray
Protection Features
  • FFP1 respirator certified in accordance with EN 149:2001+A1:2009
  • D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
  • Not reusable
Comfort Features
  • Soft material edges for increased wearer comfort
  • Seamless headband for a comfortable, secure fit
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (D marked)
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type Particulate
Material Polypropylene, Polyester
Product Type Particulate Respirator
Size Standard
With Exhalation Valve No
Filter Efficiency FFP1
Sterility Non-sterile