missing translation for 'onlineSavingsMsg'
Learn More

Pregnanetriolone, TRC

Product Code. 30008855
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008855 10 mg 10mg
30010276 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008855 Supplier Toronto Research Chemicals Supplier No. TRCP70522010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-99-6
Molecular Formula C21H34O4
Molecular Weight (g/mol) 350.49
Synonym 3α,17,20α-Trihydroxy-5β-pregnan-11-one, 11-keto-Pregnane-3α,17,20α-triol, 3α,17α,20α-Trihydroxy-5β-pregnan-11-one, 5β-Pregnan-3α,17α,20α-triol-11-one, 5β-Pregnane-3α,17α,20α-triol-11-one, NSC 79097
IUPAC Name (3R,5R,8S,9S,10S,13S,14S,17R)-3,17-dihydroxy-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,5,6,7,8,9,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-11-one
SMILES C[C@H](O)[C@@]1(O)CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H34O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h12-16,18,22-23,25H,4-11H2,1-3H3/t12-,13+,14+,15-,16-,18+,19-,20-,21-/m0/s1
Formula Weight 350.2457
Chemical Name or Material Pregnanetriolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.