missing translation for 'onlineSavingsMsg'
Learn More

Preladenant, MedChemExpress

Product Code. 16460577
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460577

Brand: Medchem Express HY10889

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Preladenant is a potent and competitive antagonist of the human adenosine A2A receptor with a Ki of 1.1 nM and has over 1000-fold selectivity over other adenosine receptors.

Preladenant is a potent and competitive antagonist of the human adenosine A2A receptor with a Ki of 1.1 nM and has over 1000-fold selectivity over other adenosine receptors.

Specifications

Chemical Name or Material Preladenant
CAS 377727-87-2
Color White
Purity Grade Notes Research
Molecular Formula C25H29N9O3
Quantity 50 mg
Synonym SCH-420814
Solubility Information DMSO : 5 mg/mL (9.93 mM; Need ultrasonic)
SMILES NC1=NC(N(CCN2CCN(C3=CC=C(OCCOC)C=C3)CC2)N=C4)=C4C5=NC(C6=CC=CO6)=NN51
Molecular Weight (g/mol) 503.56
Formula Weight 503.56
Percent Purity 98.5%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.