missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PREPL Recombinant Rabbit Monoclonal Antibody (24GB990)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557639
This item is not returnable.
View return policy
Description
PREPL Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
Specifications
| PREPL | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Prepl | |
| 2810457N15Rik; 9530014L06Rik; D030028O16Rik; KIAA0436; mKIAA0436; Prepl; prolyl endopeptidase; prolyl endopeptidase-like; Prolylendopeptidase-like; putative prolyl oligopeptidase; RGD1310143 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 213760, 298771, 9581 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB990 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4J6C6, Q5HZA6, Q8C167 | |
| Prepl | |
| A synthesized peptide derived from human PREPL (aa 50-100). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction