missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Preprotachykinin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-82555-25ul
This item is not returnable.
View return policy
Description
Preprotachykinin-1 Polyclonal antibody specifically detects Preprotachykinin-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Preprotachykinin-1 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Hs.2563, NK2, NKNA, NPK, PPT, protachykinin-1, TAC1, TAC2 | |
| This antibody was developed against Preprotachykinin-1 Recombinant Protein corresponding to amino acids: WSDWYDSDQIKEELPEPFEHLLQRIARRPKPQQFFGLMGKRDADSSIEKQVALLKALYGHGQISHKRHKTDSFVGLMGKRALNSVAYERSAMQNYERRR | |
| 25 μL | |
| Growth and Development, Neuroscience | |
| 6863 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction