missing translation for 'onlineSavingsMsg'
Learn More

Prim-O-glucosylcimifugin, MedChemExpress

Product Code. 16469048
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Packungsgröße:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
16469048 10 mg 10mg
16441468 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16469048 Lieferant Medchem Express Lieferanten-Nr. HYN0635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Prim-O-glucosylcimifugin exerts anti-inflammatory effects through the inhibition of iNOS and COX-2 expression by through regulating JAK2/STAT3 signaling.

Prim-O-glucosylcimifugin exerts anti-inflammatory effects through the inhibition of iNOS and COX-2 expression by through regulating JAK2/STAT3 signaling.

Specifications

Chemical Name or Material Prim-O-glucosylcimifugin
CAS 80681-45-4
Color White
Purity Grade Notes Research
Molecular Formula C22H28O11
Quantity 10 mg
Solubility Information DMSO : ≥ 150 mg/mL (320.20 mM)
SMILES COC1=C(C[C@@H](C(C)(O)C)O2)C2=CC(OC(CO[C@@H]([C@@H]([C@@H](O)[C@@H]3O)O)O[C@@H]3CO)=C4)=C1C4=O
Molecular Weight (g/mol) 468.45
Formula Weight 468.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.