missing translation for 'onlineSavingsMsg'
Learn More

Primaquine diphosphate, MedChemExpress

Product Code. 16465278
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465278 10 g 10g
16427997 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16465278 Supplier Medchem Express Supplier No. HY12651
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Primaquine Diphosphate (Primaquine phosphate), an 8-aminoquinoline, exerts a broad spectrum of activities against various stages of parasitic malaria. Primaquine Diphosphate remains as the only agent that destroys late hepatic stages and latent tissue forms of Plasmodium vivax and Plasmodium ovale.

Primaquine Diphosphate (Primaquine phosphate), an 8-aminoquinoline, exerts a broad spectrum of activities against various stages of parasitic malaria. Primaquine Diphosphate remains as the only agent that destroys late hepatic stages and latent tissue forms of Plasmodium vivax and Plasmodium ovale.

Specifications

Chemical Name or Material Primaquine diphosphate
Physical Form Powder
CAS 63-45-6
Color Orange
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C15H27N3O9P2
For Use With (Application) COVID-19-immunoregulation
Synonym Primaquine phosphatePrimaquine bisphosphate
Solubility Information DMSO : 250 mg/mL (549.04 mM; Need ultrasonic) ∣H2O : 25 mg/mL (54.90 mM; ultrasonic and warming and heat to 60°C)
SMILES CC(NC1=C2N=CC=CC2=CC(OC)=C1)CCCN.O=P(O)(O)O.O=P(O)(O)O
Molecular Weight (g/mol) 455.34
Formula Weight 455.34
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.