missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKAR2A Recombinant Rabbit Monoclonal Antibody (24GB1265)

Rabbit Recombinant Monoclonal Antibody
198.00 € - 564.00 €
Specifications
| Antigen | PRKAR2A |
|---|---|
| Clone | 24GB1265 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275550
![]() |
Invitrogen™
MA557622 |
100 μL |
564.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275286
|
Invitrogen™
MA557623 |
20 μL |
198.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRKAR2A Recombinant Monoclonal Antibody for Western Blot, Flow
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase (AMPK), which transduces the signal through phosphorylation of different target proteins. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans. The protein encoded by this gene is one of the regulatory subunits. This subunit can be phosphorylated by the activated catalytic subunit. It may interact with various A-kinase anchoring proteins and determine the subcellular localization of AMPK.Specifications
| PRKAR2A | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| PRKAR2A | |
| 1110061A24Rik; AI317181; AI836829; cAMP-dependent protein kinase regulatory subunit RII alpha; cAMP-dependent protein kinase type II-alpha regulatory subunit; cAMP-dependent protein kinase, regulatory subunit alpha 2; MGC3606; PKR2; PRKAR2; Prkar2a; protein kinase A, RII-alpha subunit; protein kinase cAMP-dependent regulatory type II alpha; protein kinase cAMP-dependent type II regulatory subunit alpha; protein kinase, cAMP dependent regulatory, type II alpha; protein kinase, cAMP-dependent, regulatory subunit type II alpha; protein kinase, cAMP-dependent, regulatory, type 2, alpha; protein kinase, cAMP-dependent, regulatory, type II, alpha; RII(alpha); unnamed protein product | |
| PRKAR2A | |
| Primary | |
| Affinity chromatography |
| 24GB1265 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13861 | |
| 5576 | |
| A synthesized peptide derived from human PKA R2 (aa 312-404). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title