Learn More
PRKCB Mouse, anti-Human, Clone: 4B7A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68620-1-IG-20UL
Description
The PRKCB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRKCB. This antibody recognizes human, pig, rabbit, rat, mouse antigen. The PRKCB antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PRKCB | |
| 4B7A1 | |
| PKC B, PKC beta, PKCB, PRKCB, PRKCB1, PRKCB2, Protein kinase C beta type, protein kinase C, beta | |
| 20 μL | |
| 5579 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig, Rabbit, Rat, Mouse | |
| 150 kDa | |
| IgG1 |