Learn More
PRKCB Mouse, anti-Human, Clone: 4B7A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68620-1-IG-150UL
 View more versions of this product
Product Code. 18076385
  • 410.00 € / 150µL
Description
The PRKCB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRKCB. This antibody recognizes human, pig, rabbit, rat, mouse antigen. The PRKCB antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PRKCB
4B7A1
PKC B, PKC beta, PKCB, PRKCB, PRKCB1, PRKCB2, Protein kinase C beta type, protein kinase C, beta
150 μL
5579
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
500 μg/mL
Mouse
Primary
Human, Pig, Rabbit, Rat, Mouse
150 kDa
IgG1