missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKCSH Recombinant Rabbit Monoclonal Antibody (24GB1325)

Rabbit Recombinant Monoclonal Antibody
198.00 € - 564.00 €
Specifications
| Antigen | PRKCSH |
|---|---|
| Clone | 24GB1325 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275983
![]() |
Invitrogen™
MA557624 |
100 μL |
564.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276275
|
Invitrogen™
MA557625 |
20 μL |
198.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRKCSH Recombinant Monoclonal Antibody for Western Blot, Flow
SET domain-containing proteins, such as SMYD2, catalyze lysine methylation.Specifications
| PRKCSH | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| PRKCSH | |
| 80K-H; 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; AGE-R2; alpha glucosidase II, beta-subunit; carbohydrate processing enzyme of the endoplasmic reticulum; G19P1; GIIB; GLU2B; glucosidase 2 subunit beta; glucosidase II; glucosidase II beta-subunit; glucosidase II subunit beta; hepatocystin; PCLD; PKCSH; PLD1; PRKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate 80K-H; protein kinase C substrate, 80 Kda protein; VASAP-60 | |
| PRKCSH | |
| Primary | |
| Affinity chromatography |
| 24GB1325 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14314 | |
| 5589 | |
| A synthesized peptide derived from human GLU2B (aa 501-528). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title