Learn More
PRKDC Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-28534-100UL
Description
The PRKDC antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PRKDC. This antibody recognizes human antigen. The PRKDC antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| PRKDC | |
| 1000 μg/mL | |
| DNA PK catalytic subunit, DNA PKcs, DNAPK, DNPK1, HYRC, HYRC1, p350, p460, PRKDC, XRCC7 | |
| 100 μL | |
| 5591 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |