Learn More
PRKRA Mouse, anti-Human, Clone: 1D4C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68518-1-IG-150UL
 View more versions of this product
Product Code. 18065475
  • 410.00 € / 150µL
Description
The PRKRA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRKRA. This antibody recognizes human, mouse antigen. The PRKRA antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PRKRA
1D4C3
DYT16, HSD 14, HSD14, PACT, PKR associated protein X, PKR associating protein X, PRKRA, RAX
150 μL
8575
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse
150 kDa
IgG2b