missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRMT1 Recombinant Rabbit Monoclonal Antibody (24GB5045)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557627
This item is not returnable.
View return policy
Description
PRMT1 Recombinant Monoclonal Antibody for Western Blot
PRMT1 is a member of the protein arginine N-methyltransferase (PRMT) family. Arginine methylation is an irreversible post translational modification which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Increased expression of PRMT1 may play a role in various types of cancer.
Specifications
| PRMT1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRMT1 | |
| 6720434D09Rik; ANM1; arginine N-methyltransferase 1; AW214366; HCP1; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; heterogeneous nuclear ribonucleoproteins methyltransferase-like 2; histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; HMT1 hnRNP methyltransferase-like 2; HMT1 hnRNP methyltransferase-like 2 isoform 3; HMT2; Hrmt1l2; interferon receptor 1-bound protein 4; IR1B4; Mrmt1; PRMT1; protein arginine methyltransferase 1; protein arginine N-methyltransferase 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15469, 3276, 60421 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB5045 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q63009, Q99873, Q9JIF0 | |
| PRMT1 | |
| A synthesized peptide derived from human PRMT1 (aa 327-340). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction