Learn More
Invitrogen™ PRMT1 Recombinant Rabbit Monoclonal Antibody (24GB5045)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557626
 View more versions of this product
Product Code. 30276233
  • 564.00 € / 100µL
Description
PRMT1 Recombinant Monoclonal Antibody for Western Blot
PRMT1 is a member of the protein arginine N-methyltransferase (PRMT) family. Arginine methylation is an irreversible post translational modification which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Increased expression of PRMT1 may play a role in various types of cancer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRMT1
Recombinant Monoclonal
Unconjugated
PRMT1
6720434D09Rik; ANM1; arginine N-methyltransferase 1; AW214366; HCP1; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; heterogeneous nuclear ribonucleoproteins methyltransferase-like 2; histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; HMT1 hnRNP methyltransferase-like 2; HMT1 hnRNP methyltransferase-like 2 isoform 3; HMT2; Hrmt1l2; interferon receptor 1-bound protein 4; IR1B4; Mrmt1; PRMT1; protein arginine methyltransferase 1; protein arginine N-methyltransferase 1
Rabbit
Affinity chromatography
RUO
15469, 3276, 60421
-20°C
Liquid
Flow Cytometry, Western Blot
24GB5045
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q63009, Q99873, Q9JIF0
PRMT1
A synthesized peptide derived from human PRMT1 (aa 327-340).
100 μL
Primary
Human, Mouse, Rat