missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT5 Antibody (RAB-C136), DyLight 680 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20131FR
This item is not returnable.
View return policy
Description
PRMT5 Monoclonal antibody specifically detects PRMT5 in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
| PRMT5 | |
| Monoclonal | |
| DyLight 680 | |
| EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT5 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Circadian Rhythm | |
| 10419 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunofluorescence | |
| RAB-C136 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction