Learn More
Invitrogen™ PRMT5 Recombinant Rabbit Monoclonal Antibody (24GB845)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557628
 View more versions of this product
Product Code. 30275830
  • 564.00 € / 100µL
Description
PRMT5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PRMT5 (protein arginine N-methyltransferase 5) is an arginine methyltransferase that can both catalyze the formation of omega-N monomethylarginine (MMA) and symmetrical dimethylarginine (sDMA), with a preference for the formation of MMA. PRMT5 specifically mediates the symmetrical dimethylation of arginine residues in the small nuclear ribonucleoproteins Sm D1 and Sm D3; such methylation is required for the assembly and biogenesis of snRNP core particles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRMT5
Recombinant Monoclonal
Unconjugated
PRMT5
72 kDa ICln-binding protein; EC 2.1.1.125; histone-arginine N-methyltransferase PRMT5; HMT1 hnRNP methyltransferase-like 5; HRMT1L5; hypothetical protein; I79_013398; IBP72; Jak-binding protein 1; JBP1; PRMT5; protein arginine methyltransferase 5; protein arginine N-methyltransferase 5; Protein arginine N-methyltransferase 5, N-terminally processed; protein arginine N-methyltransferase 5-like; shk1 kinase-binding protein 1 homolog; Skb1; SKB1 homolog; SKB1Hs
Rabbit
Affinity chromatography
RUO
10419, 27374, 364382
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB845
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O14744, Q8CIG8
PRMT5
A synthesized peptide derived from human PRMT5 (aa 40-92).
100 μL
Primary
Human, Mouse, Rat