Learn More
Invitrogen™ PRMT6 Recombinant Rabbit Monoclonal Antibody (24GB1585)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557630
 View more versions of this product
Product Code. 30274854
  • 564.00 € / 100µL
Description
PRMT6 Recombinant Monoclonal Antibody for Western Blot, Flow
Protein arginine N-methyltransferase 6 (PRMT6) PRMT6 demonstrates type I PRMT activity, capable of forming both omega-N monomethylarginine and asymmetric dimethylarginine derivatives on the recombinant glycine- and arginine-rich substrates. cts as a transcriptional coactivator of a number of steroid hormone receptors including ESR1, ESR2, PGR and NR3C1. Promotes fasting-induced transcriptional activation of the gluconeogenic program through methylation of the CRTC2 transcription coactivator. May play a role in innate immunity against HIV-1 in case of infection by methylating and impairing the function of various HIV-1 proteins such as Tat, Rev and Nucleocapsid protein p7.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRMT6
Recombinant Monoclonal
Unconjugated
PRMT6
AW124876; BB233495; FLJ10559; FLJ51477; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6; Histone-arginine N-methyltransferase PRMT6; HMT1 hnRNP methyltransferase-like 6; Hrmt1l6; PRMT6; PRMT-6; protein arginine methyltransferase 6; protein arginine N-methyltransferase 6
Rabbit
Affinity chromatography
RUO
295384, 55170, 99890
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1585
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6NZB1, Q96LA8
PRMT6
A synthesized peptide derived from human PRMT6 (aa 203-226).
100 μL
Primary
Human, Mouse, Rat