Learn More
Invitrogen™ pro-GDF3 Monoclonal Antibody (356613)
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA561341
Product Code. 30534943
  • 429.00 € / 100µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Pro-GDF3, or pro-growth differentiation factor 3, is the precursor form of GDF3, a member of the transforming growth factor-beta (TGF-beta) superfamily. GDF3 is involved in various biological processes including embryonic development, cell differentiation, and regulation of adipose tissue homeostasis. Pro-GDF3 undergoes proteolytic processing to become the mature, active form that can then bind to its receptors and initiate signaling pathways. GDF3 plays a critical role in early embryogenesis, particularly in the formation of the mesoderm and endoderm, and in maintaining the pluripotency of embryonic stem cells. Additionally, GDF3 has been implicated in metabolic regulation and in the pathogenesis of obesity and type 2 diabetes. Abnormal expression or mutations in the GDF3 gene can lead to developmental disorders and contribute to disease states. As a result, pro-GDF3 and its mature form are of significant interest in developmental biology, regenerative medicine, and metabolic disease research, offering potential targets for therapeutic intervention.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
pro-GDF3
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Mouse myeloma cell line NS0-derived recombinant mouse GDF-3 Propeptide, Ser23-Lys250.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
356613
Unconjugated
Rat
Protein A/G
RUO
Mouse
Antibody
IgG2a