missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pro-IL-33 Monoclonal Antibody (518017)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561510
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Pro-IL-33 is the precursor form of interleukin-33 (IL-33), a cytokine that plays an essential role in the immune system, particularly in the regulation of inflammatory and immune responses. IL-33 is a member of the IL-1 family and is produced by various cell types, including epithelial cells, endothelial cells, and fibroblasts. In its precursor form, pro-IL-33 is biologically active and can be cleaved by caspases or other proteases to generate the mature, active cytokine. IL-33 functions by binding to its receptor, ST2 (IL1RL1), on the surface of target cells, which include a variety of immune cells such as Th2 cells, mast cells, and group 2 innate lymphoid cells (ILC2s). This interaction leads to the activation of signaling pathways that promote the production of Th2-associated cytokines, contributing to the immune response against infections and playing a role in allergic inflammation. Elevated levels of IL-33 have been associated with various inflammatory and autoimmune diseases, including asthma, rheumatoid arthritis, and inflammatory bowel disease (IBD). Due to its significant role in modulating immune responses, pro-IL-33 is a critical target for research aimed at developing new treatments for inflammatory and autoimmune conditions.
Specifications
| pro-IL-33 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| E. coli-derived recombinant mouse IL-33 Propeptide, Met1-Leu108. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 518017 | |
| Unconjugated | |
| Rat | |
| Protein A/G | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction