missing translation for 'onlineSavingsMsg'
Learn More

Procaine hydrochloride, MedChemExpress

Product Code. 16460988
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460988 5 g 5g
16422229 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460988 Supplier Medchem Express Supplier No. HYB0546A
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Procaine hydrochloride is a DNA-demethylating agent. Procaine hydrochloride acts through multiple targets and has a slow onset and a short duration of action.

Procaine hydrochloride is a DNA-demethylating agent. Procaine hydrochloride acts through multiple targets and has a slow onset and a short duration of action.

Specifications

Chemical Name or Material Procaine hydrochloride
Physical Form Solid
CAS 51-05-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C13H21ClN2O2
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : ≥ 50 mg/mL (183.30 mM) ∣DMSO : 50 mg/mL (183.30 mM; Need ultrasonic)
SMILES O=C(OCCN(CC)CC)C1=CC=C(N)C=C1.Cl
Molecular Weight (g/mol) 272.77
Formula Weight 272.77
Percent Purity 99.94%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.