missing translation for 'onlineSavingsMsg'
Learn More

Procarbazine Hydrochloride, MedChemExpress

Product Code. 16476296
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476296 100 mg 100mg
16471696 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16476296 Supplier Medchem Express Supplier No. HY13733
 Restricted Item
Estimated Shipment: 01-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress Procarbazine Hydrochloride is an orally active alkylating agent, with anticancer activity. Procarbazine Hydrochloride can be used in Hodgkin's disease research.

Procarbazine Hydrochloride is an orally active alkylating agent, with anticancer activity. Procarbazine Hydrochloride can be used in Hodgkin's disease research.

Specifica

Chemical Name or Material Procarbazine Hydrochloride
Physical Form Solid
CAS 366-70-1
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H20ClN3O
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : 100 mg/mL (387.96 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (64.67 mM; Need ultrasonic)
SMILES O=C(NC(C)C)C1=CC=C(CNNC)C=C1.Cl
Molecular Weight (g/mol) 257.76
Formula Weight 257.76
Percent Purity 95.0%
Grade Research
Vedi altri risultati Mostra meno risultati

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.