missing translation for 'onlineSavingsMsg'
Learn More

Procodazole, MedChemExpress

Product Code. 16462246
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 16462246

Brand: Medchem Express HYB1056

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Procodazole is a non-specific active immunoprotective agent against viral and bacterial infections, used as a potentiator.

Procodazole is a non-specific active immunoprotective agent against viral and bacterial infections, used as a potentiator.

Specifications

Chemical Name or Material Procodazole
Physical Form Solid
CAS 23249-97-0
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C10H10N2O2
For Use With (Application) COVID-19-anti-virus
Synonym Propazol 2-Benzimidazolepropionic acid
Solubility Information DMSO : 50 mg/mL (262.88 mM; Need ultrasonic)
SMILES O=C(O)CCC1=NC2=CC=CC=C2N1
Molecular Weight (g/mol) 190.2
Formula Weight 190.2
Percent Purity 99.88%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.