missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ Product Manual

Product Code. 15339624 Shop All Velp Products
Click to view available options
For Use With (Equipment):
COEX
CSF6
FIWE Series Raw Fiber Extractors
GDE
HU6
JP
NDA 701 and 702
OXITEST
SER 148 Series Solvent Extractors
SMS
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15339624

Brand: Velp Scientifica™ A00000253

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Velp Scientifica™ Product Manual are for use with a wide range of products.

  • Inlcudes one manual for the desiered product
  • For use with a wide range of Velp Scientifica™ products

Specifications

Format Manual
For Use With (Equipment) JP
Product Type Operating Instructions
Quantity 1/Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.