missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neon™ Transfection System Starter Pack

Animal origin-free, serum-free and compatible with transfection-grade nucleic acid purification kits, siRNA vectors and DNA vectors

Brand:  Invitrogen™ MPK5000S

10355.77 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.




Additional Details : Weight : 6.00000kg

Product Code. 10090314

  • 11460.00 € / Each
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Neon Electroporation Device, Pipette, Pipette Station, USB thumbdrive, Manual, Power Cord, one Neon Transfection System 100μL Kit (192 reactions) and one Neon Transfection System 10μL Kit (192 reactions)

Description

Description

Neon Transfection System enables fast and efficient delivery of nucleic acids into all mammalian cell types, including primary, stem, and difficult-to-transfect cells. The flexible and open system allows the user to perform high-quality transfections using optimized or user-defined protocols in three simple steps with as few as 2 x 104 cells per reaction and a reaction time of just 1 second. A novel reaction chamber provides a dramatic increase in transfection efficiency and cell viability.

  • Efficient:up to 90% in many cell types, including difficult-to-transfect cells, primary, and stem cells
  • Flexible:easily transfect from 2 x 104 cells to 6 x 106 cells per reaction
  • Simple:easy to use, with a single reagent kit for all cell types
  • Versatile:open system allows electroporation parameters to be optimized freely
  • One-of-a-kind electroporation:Unlike standard cuvette-based electroporation chambers, the Neon system uses a patented, biologically compatible pipette tip chamber that generates a more uniform electric field
  • This design allows better maintenance of physiological conditions, resulting in very high cell survival compared to conventional electroporation
  • Plug-n-play out of the box:The transfection process couldn't be simpler
  • Simplified transfection kit for all cell types
  • Voltage limit: 0.5 to 2.5KV

Adenoviral shRNA, Cell Culture, Lentiviral shRNA & miRNA, Plasmid Transfection, RNAi, RNAi Transfection, RNAi, Epigenetics & Noncoding RNA Research, Retroviral RNAi, Stem Cell & Primary Cell Transfections, Synthetic siRNA Transfection, Transfection, Vector-Based RNAi, siRNA

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

3.0-6.0 A
The Neon™ Transfection System Starter pack comes with:
• 1 Neon™ Electroporation Device
• 1 Pipette
• 1 Pipette Station
• 1 USB thumbdrive
• 1 Manual
• 1 Power Cord

Also included are one Neon™ Transfection System 100 μl Kit (192 reactions) and one Neon™ Transfection System 10 μl Kit (192 reactions). Store the buffer boxes 4°C and the tips and tubes boxes at room temperature.
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Neon™ Transfection System
0.5-2.5 kV
Transfection System Starter Pack
1 Pack
Yes
Neon™ Transfection System
300 W
Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum
Stem Cells, Primary Cells, Hard-to-Transfect Cells
Plasmid DNA, Synthetic siRNA
Transfection
Neon™
Transfection Grade
1 sec
2x10^4 - 6 x10^8 cells; Varies by cell type
100-240 VAC
10μl and 100μl
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.