missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase with Gold Buffer and MgCl2

Chemically modified form of AmpliTaq DNA Polymerase requiring thermal activation

Brand:  Applied Biosystems™ 4317742

14483.05 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.




Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg

 View more versions of this product

Product Code. 10640796

  • 17090.00 € / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Hot-start activation
The modified enzyme is provided in an inactive state. Upon thermal activation, the modifier is permanently released, regenerating active enzyme. The resulting hot-start PCR amplification provides increased sensitivity, specificity, and yield over conventional PCR techniques.

Higher specificity, higher yield
AmpliTaq Gold DNA Polymerase can be activated partially or completely in a pre-PCR heat step, or can be allowed to activate slowly in a time-released manner during the denaturation steps of thermal cycling. With or without a limited up-front heat activation step, active enzyme is released slowly during thermal cycling to match template concentration and increase specificity. The yield of specific product increases because reactants are not wasted in the formation of unintended products. Because AmpliTaq Gold DNA Polymerase is a chemical hot-start enzyme, there is no worry of biological contamination. GeneAmp 10X PCR Gold Buffer is formulated to provide flexible, efficient activation of AmpliTaq Gold DNA Polymerase, resulting in a highly specific, robust PCR amplification. The ionic strength and the pH of GeneAmp 10X PCR Gold Buffer have been optimized to provide a wider activation temperature and time range when used in conjunction with AmpliTaq Gold DNA Polymerase.

AmpliTaq Gold 360 DNA Polymerase for superior performance
Also available is AmpliTaq Gold 360 DNA Polymerase for even better PCR performance. Compared to AmpliTaq Gold DNA Polymerase, AmpliTaq Gold 360 DNA Polymerase is purified by an additional proprietary separation process to eliminate contaminating bacterial DNA sequences from the enzyme preparation. When used with the enhanced 360 Gold Buffer, this ultra-pure enzyme, in addition to its hot-start capabilities, reduces false positive results, amplifies low-level target sequences, and promotes the amplification of a variety of templates. Supplied 360 GC Enhancer works through challenging GC-rich templates.

  • Time-released thermal activation improves sensitivity in low copy number amplifications
  • Successful multiplex PCR saves time and reagents
  • Includes GeneAmp 10X PCR Gold Buffer with MgCl2
  • Automated chemical hot-start enzyme for increased specificity, sensitivity, and yield

Hot Start PCR, PCR, PCR and Real-Time PCR

Specifications

Specifications

Contains 5 boxes total, each of which contains the following:
• 5 tubes (200 μl at 5 U/μl) of AmpliTaq Gold™ DNA Polymerase (250 units total per tube)
• 20 tubes (1.5 mL) of GeneAmp™ 10X Gold Buffer (150 mM Tris-HCl, pH 8.0, 500 mM KCl)
• 20 tubes (1.5 mL) of 25 mM magnesium chloride.

Store at -15 to -30°C.
Tube
AmpliTaq Gold DNA Polymerase
5' - 3'
DNA Polymerase
5 U/μL
Built-In Hot Start
3'-A
5 kb or less
Primer-probe
Low
Standard
AmpliTaq Gold™
25,000 Units
1 X
20000
Separate Components
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.