Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit
Description
- Charges TR-630 standard battery or TR-632 high capacity battery in less than 4 hours
- Batteries may be stored on the charger, fully charged, ready for immediate use
Specifications
Specifications
| Product Type | Single Station Battery Charger Kit |
| For Use With (Equipment) | 3M Versaflo Powered Air Turbo TR-600 Series |
| Includes | TR-640 Cradle, Power Supply and Lead |
| Model | TR-641E |