missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TOM1L2 Protein

A cDNA sequence encoding the TOM1L2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13781-1mg


Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924689

  • 7540.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TOM1L2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSEFMEFLL GNPFSTPVGQ CLEKATDGSL QSEDWTLNME ICDIINETEE GPKDAIRALK KRLNGNRNYR EVMLALTVLE TCVKNCGHRF HILVANRDFI DSVLVKIISP KNNPPTIVQD KVLALIQAWA DAFRSSPDLT GVVHIYEELK RKGVEFPMADLDALSPIHTP QRSVPEVDPA ATMPRSQSQQ RTSAGSYSSP PPAPYSAPQA PALSVTGPIT ANSEQIARLR SELDVVRGNT KVMSEMLTEM VPGQEDSSDL ELLQELNRTC RAMQQRIVEL ISRVSNEEVT EELLHVNDDL NNVFLRYERF ERYRSGRSVQ NASNGVLNEV TEDNLIDLGP GSPAVVSPMV GNTAPPSSLS SQLAGLDLGT ESVSGTLSSL QQCNPRDGFD MFAQTRGNSL AEQRKTVTYE DPQAVGGLAS ALDNRKQSSE GIPVAQPSVM DDIEVWLRTD LKGDDLEEGV TSEEFDKFLE ERAKAAEMVP DLPSPPMEAP APASNPSGRK KPERSEDALF AL
Greater than 80.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TOM1L2 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers