missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other C. Albicans Met6 Protein

A cDNA sequence encoding the C. Albicans Met6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11218-1mg


Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956518

  • 5800.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

C. Albicans Met6 Protein
Research Use Only
Recombinant Protein
Baculovirus
C. Albicans Met6 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers