missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Productainer™ 3D BioProcess Container (BPC), Aegis™ 5-14 film, two ports, MPCs

Product Code. 17890587 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
Unit Size:
Pack of 10
Pack of 5
This item is not returnable. View return policy

Product Code. 17890587

Brand: Thermo Scientific™ SH30964.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific Productainer 3D BioProcess Containers (BPCs) are used to package, store, and transport media, buffers, water for injection (WFI), process liquids, and biological products.

Productainer BPCs are constructed with Aegis 5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainer BPCs are suitable for a wide array of bioprocess applications. For optimal fit, form, and function, Productainer BPCs can be customized using the industry's largest component library.

Highlights:
• Film optimized for flexibility and durability—superior stress resistance, puncture resistance, and physical barrier properties to ensure stability of contents
• Gamma-irradiation sterilized
• Squared tube design with polyethylene ports located on the top of the BPC for easy access
• Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
• Customizable via the industry's largest library of qualified components

Recommended for:
• Harvest—collection and storage of bulk product from bioreactors
• Storage—sterile storage of media, buffers, and process intermediates
• Separation—feeding and receiving of sterile liquids from separation systems
• Purification—buffer feeding to purification system, fraction collection and storage
• Filling—sterile reservoir for filling systems
• Transport and shipping—bulk sterile liquids, buffers, media and process intermediates within and between facilities

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 100 L
Description Top Dispense
Film Description Aegis 5-14
Port Detail 2 ports
Quantity 100 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.