Learn More
Thermo Scientific™ Productainer™ BioProcess Container (BPC), 100L with 2 Top and 1 Bottom Ports, 3D Single-Use BPC
Shop All Thermo Scientific Products

Beschreibung
Using Thermo Scientific™ Productainer™ BPC bag with 2 top and 1 bottom ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.
Highlights:
- Three sizes - 50, 100 and 200L
- Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
- Gamma-irradiation sterilized
- Squared tube design with polyethylene ports located on the top of the BPC for easy access
- Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
- Customizable via the industry's largest library of qualified components
Recommended for:
Harvest - Collection and storage of bulk product from bioreactors
Storage - Sterile storage of media, buffers and process intermediates
Separation - Feeding and receiving of sterile liquids from separation systems
Purification - Buffer feeding to purification system, fraction collection and storage
Filling - Sterile reservoir for filling systems
Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities
Spezifikation
Spezifikation
| Capacity (Metric) | 100 L |
| Material | Polyethylene |
| Outer Packaging | Packages placed in strong cardboard cartons with product and lot identification |
| Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
| Quantity | 100 L |
| For Use With (Equipment) | SV50517.05 |
| Sterilization Method | Gamma Irradiated |
| Tubing Line 1 | Line 1. 19.1 mm (0.75 in.) tri-clamp, polyethylene, Tubing: C-Flex, length: 45.7 cm (18 in.) ID x OD: 12.7 x 19.1 mm (0.5 in. x 0.75 in.) Top Port |
| Tubing Line 3 | Line 3. 19.1 mm (0.75 in.) tri-clamp, polyethylene, Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 12.7 x 19.1 mm (0.5 in. x 0.75 in.) Bottom Port |
| Tubing Line 2 | Line 2. 19.1 mm (0.75 in.) tri-clamp, polyethylene, Tubing: C-Flex, length: 45.7 cm (18 in.) ID x OD: 12.7 x 19.1 mm (0.5 in. x 0.75 in.) Top Port |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.