Learn More
Thermo Scientific™ Productainer™ BioProcess Container (BPC), 200L with 3 Ports, 2D BPC
Shop All Thermo Scientific Products

Description
The Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 200 L 2D pillow design is composed of CX5-14 single web, multi-layer plastic film and includes 3 panel ports.
Features:
- Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
- All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.
Applications of the Thermo Scientific Labtainer BPC include:
- Harvest from, feed into, and collect samples from bioreactors and fermentors.
- Buffer, culture media, and process liquid preparation and storage.
- Chromatography feed and fraction collection.
- Harvest, storage and transport of bulk drug product and bulk drug precursors.
Customize standard BPC systems for optimized single-use technology
Choose from the industry's largest component library – with over 20,000 unique components that include:
- Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
- Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
- Filters—used for bioburden reduction; sterilization grade filters and vent filters.
- Tubing—thermoplastic elastomers, platinum-cured silicone.
- Sample ports—septum-style and needle-free ports.
- Clamps—sliders, pinch, and crimp bands.
Specifications
Specifications
| Green Features | ISCC PLUS |
| Capacity (Metric) | 200 L |
| Film Description | CX5-14 |
| Port Detail | Panel Ports: 2 x 0.38 in. 1 x 0.25 in. |
| For Use With (Equipment) | HyPerforma Flat-bottom LLDPE Drum - SV50076.04 |
| Dimensions | Chamber: 94 cm x 142 cm (37.5 in. x 56 in.) |
| Material | Polyethylene |
| Outer Packaging | Packages placed in strong cardboard cartons with product and lot identification |
| Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
| Quantity | 1 |
| Show More |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.