missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Profilin 1 Antibody (CL3524) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59778MFV450
This item is not returnable.
View return policy
Description
Profilin 1 Monoclonal antibody specifically detects Profilin 1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out,KnockDown
Specifications
| Profilin 1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| profilin 1, Profilin I, profilin-1 | |
| This antibody was developed using a recombinant protein derived from P07737, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
| 5216 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out, KnockDown | |
| CL3524 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction