missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Profilin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00301-100ul
This item is not returnable.
View return policy
Description
Profilin 2 Polyclonal antibody specifically detects Profilin 2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications
| Profilin 2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:2500-1:10000 | |
| D3S1319E, PFL, profilin 2, Profilin II, profilin-2 | |
| Produced in rabbits immunized with purified, recombinant Human Profilin 2 (Accession#: AAH18049.1; Met1-Phe140) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Signal Transduction, Stem Cell Markers | |
| 5217 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction