missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R B/NR3C3 Antibody (CL8721) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21167MFV450
This item is not returnable.
View return policy
Description
Progesterone R B/NR3C3 Monoclonal antibody specifically detects Progesterone R B/NR3C3 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Progesterone R B/NR3C3 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5241 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL8721 | |
| 50mM Sodium Borate | |
| This antibody was developed using a recombinant protein derived from P06401, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction