missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proinsulin Antibody (CCI-17), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-73013
This item is not returnable.
View return policy
Description
Proinsulin Monoclonal specifically detects Proinsulin in Human, Rat samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
Specifications
| Proinsulin | |
| Monoclonal | |
| Unconjugated | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3630 | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA | |
| CCI-17 | |
| Western Blot, ELISA 1:100-1:2000, Sandwich ELISA 1:100-1:2000 | |
| INS | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragment of rat proinsulin conjugated with a carrier protein. | |
| 0.2 mg | |
| Primary | |
| Rat proinsulin. NB100-73013 binds specifically to proinsulin and does not bind to insulin or C-peptide. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction