missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prokineticin 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94101-0.02ml
This item is not returnable.
View return policy
Description
Prokineticin 2 Polyclonal antibody specifically detects Prokineticin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Prokineticin 2 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin | |
| BV8Protein Bv8 homolog, KAL4, MIT1, PK2prokineticin-2, prokineticin 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-129 of human PROK2 (NP_001119600.1). LTPRAGDAAVITGACDKDSQCGGGMCCAVSIWVKSIRICTPMGKLGDSCHPLTRKNNFGNGRQERRKRKRSKRKKEVPFFGRRMHHTCPCLPGLACLRTSFNRFICLAQK | |
| 0.02 mL | |
| Angiogenesis | |
| 60675 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction