Learn More
Prolactin Antibody (CL6553) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76503IR
Product Code. 30289710
  • 748.00 € / 0.10mL
Description
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Prolactin
Monoclonal
DyLight 755
prolactin
This antibody was developed using a recombinant protein derived from P01236, with the exact immunogen sequence remaining proprietary.
0.1 mL
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction
5617
Store at 4⪚C in the dark.
IgG2a
Western Blot
CL6553
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified