Learn More
Prolactin Antibody (CL6559) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76505JF549
Product Code. 30290568
  • 748.00 € / 0.10mL
Description
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Prolactin
Monoclonal
Janelia Fluor 549
prolactin
This antibody was developed using a recombinant protein derived from P01236, with the exact immunogen sequence remaining proprietary.
0.1 mL
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction
5617
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL6559
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified