missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin R Antibody (SPM213), Novus Biologicals™
Mouse Monoclonal Antibody
265.00 € - 474.00 €
Specifications
| Antigen | Prolactin R |
|---|---|
| Clone | SPM213 |
| Dilution | Western Blot 0.25-0.5ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18111824
|
Novus Biologicals
NBP2-32898-0.02MG |
0.02 mg |
265.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18157724
|
Novus Biologicals
NBP2-32898-0.1MG |
0.1 mg |
474.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prolactin R Monoclonal specifically detects Prolactin R in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Prolactin R | |
| Western Blot 0.25-0.5ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P16471 | |
| 5618 | |
| Semi-purified human prolactin receptor | |
| Primary | |
| Store at 4C. | |
| 70 kDa |
| SPM213 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein | |
| PRLR | |
| IgG1 κ | |
| Protein A purified | |
| It recognizes a protein of 70kDa, identified as prolactin receptor. Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor), which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title