missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolyl endopeptidase-like Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93305-0.02ml
This item is not returnable.
View return policy
Description
Prolyl endopeptidase-like Polyclonal antibody specifically detects Prolyl endopeptidase-like in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Prolyl endopeptidase-like | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.83, KIAA0436FLJ16627, prolyl endopeptidase-like, Prolylendopeptidase-like, putative prolyl oligopeptidase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 578-727 of human PREPL (NP_006027.2). AVTLEAPFLDVLNTMMDTTLPLTLEELEEWGNPSSDEKHKNYIKRYCPYQNIKPQHYPSIHITAYENDERVPLKGIVSYTEKLKEAIAEHAKDTGEGYQTPNIILDIQPGGNHVIEDSHKKITAQIKFLYEELGLDSTSVFEDLKKYLKF | |
| 0.02 mL | |
| Cell Biology | |
| 9581 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction